Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40 Ligand/TNFSF4 Rabbit anti-Human, Clone: R4930 (Oxelumab), Novus Biologicals™
Rabbit Monoclonal Antibody
$229.00 - $516.00
Specifications
Antigen | OX40 Ligand/TNFSF4 |
---|---|
Clone | R4930 (Oxelumab) |
Dilution | Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready |
Applications | Flow Cytometry, Blocking Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25268725
|
Novus Biologicals
NBP2526870.025MG |
0.025 mg |
Each for $229.00
|
|
NBP25268702
|
Novus Biologicals
NBP2526870.2MG |
0.2 mg |
Each for $516.00
|
|
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
OX40 Ligand/TNFSF4 | |
Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
TNFSF4 | |
IgG | |
Protein A or G purified | |
Binds to human OX40L. |
R4930 (Oxelumab) | |
Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Immunology | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
7292 | |
Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title