Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40/TNFRSF4 Mouse anti-Feline, Biotin, Clone: 7D6, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064375B
Description
OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Feline samples. It is validated for Western Blot, Flow CytometrySpecifications
OX40/TNFRSF4 | |
Monoclonal | |
Biotin | |
ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen | |
CHO-derived feline CD134-Fc fusion protein. | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction | |
7293 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
7D6 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Feline | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction