Learn More
OX40/TNFRSF4 Mouse anti-Human, Alexa Fluor 405, Clone: 977974, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB10543V100UG
Catalog No. FB10543V100
Add to cart
This item is not returnable. View return policy
Description
Description
OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
OX40/TNFRSF4
977974
Flow Cytometry 0.25-1 μg/106 cells
P43489
ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen
Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4, Leu29-Ala216, Accession No. P43489
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human OX40/TNFRSF4 in direct ELISAs
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Conjugated, Purified, Supernatant
Mouse
IgG2b
100 μg
Primary
7293.0
Human