Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OX40/TNFRSF4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen OX40/TNFRSF4
Immunogen Synthetic peptides corresponding to TNFRSF4 (tumor necrosis factor receptor superfamily, member 4) The peptide sequence was selected from the middle region of TNFRSF4. Peptide sequence GKHTLQPASNSSDAICEDRDPPATQPQETQGPPARPITVQPTEAWPRTSQ.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16915620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16915620UL
20ul Each for $149.00
Add to cart
 
NBP169156 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169156
100 ul Each for $329.00
Add to cart
 
Description

Description

OX40/TNFRSF4 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

OX40/TNFRSF4
Rabbit
Polyclonal
Unconjugated
7293
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to TNFRSF4 (tumor necrosis factor receptor superfamily, member 4) The peptide sequence was selected from the middle region of TNFRSF4. Peptide sequence GKHTLQPASNSSDAICEDRDPPATQPQETQGPPARPITVQPTEAWPRTSQ.
Primary
IgG
ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit