Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OXA1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100165
Description
Antibody detects endogenous levels of total OXA1L.
OXA1L is required for the insertion of integral membrane proteins into the mitochondrial inner membrane. It is essential for the activity and assembly of cytochrome oxidase and is required for the correct biogenesis of ATP synthase and complex I in mitochondria.
Specifications
OXA1L | |
Polyclonal | |
Unconjugated | |
OXA1L | |
1810020M02Rik; AI894287; Hsa; hypothetical protein LOC520335; LOW QUALITY PROTEIN: mitochondrial inner membrane protein OXA1L; mitochondrial inner membrane protein OXA1L; OXA1; OXA1Hs; Oxa1l; OXA1L mitochondrial inner membrane protein; OXA1L, mitochondrial inner membrane protein; OXA1-like protein; oxidase (cytochrome c) assembly 1-like; oxidase assembly 1-like; oxidase assembly 1-like protein; zgc:163091; zgc:171949 | |
Rabbit | |
Affinity chromatography | |
RUO | |
5018, 69089, 691393 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15070, Q8BGA9 | |
OXA1L | |
A synthesized peptide derived from human OXA1L(Accession Q15070), corresponding to amino acid residues K367-I417. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction