Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic acid, 98%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
144-62-7
C2H2O4
90.034
MFCD00002573
MUBZPKHOEPUJKR-UHFFFAOYSA-N
ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico
971
CHEBI:16995
oxalic acid
C(=O)(C(=O)O)O
144-62-7
C2H2O4
90.034
MFCD00002573
MUBZPKHOEPUJKR-UHFFFAOYSA-N
ethanedioic acid,aktisal,aquisal,oxiric acid,oxalate,oxalsaeure,oxaalzuur,kyselina stavelova,acide oxalique,acido ossalico
971
CHEBI:16995
oxalic acid
C(=O)(C(=O)O)O
Assay Percent Range 98%
Beilstein 02, 502
Merck Index 15, 7010
Density 1.9000g/mL
Formula Weight 90.04
Physical Form Crystalline Powder
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.9
Solubility Solubility in water: 90g/L (20°C). Other solubilities: very soluble in ethanol, soluble in glycerol, slightly soluble in ether, insoluble in benzene, chloroform and petroleum, ether
Sublimation Point 101.0 °C
Melting Point 189.0°C to 191.0°C
Color White
Quantity 250g
Flash Point >93.4°C
Chemical Name or Material Oxalic acid
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger