Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic acid bis(cyclohexylidenehydrazide), 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
370-81-0
C14H22N4O2
278.356
MFCD00001659
DSRJIHMZAQEUJV-UHFFFAOYSA-N
cuprizone,cuprizane,biscyclohexanone oxaldihydrazone,ethanedioic acid, bis cyclohexylidenehydrazide,oxalic acid bis cyclohexylidenehydrazide,biscyclohexanone oxalyldihydrazone,bis cyclohexanone oxaldihydrazone,cuprizon,unii-5n16u7e0ao,oxalic acid, bis cyclohexylidenehydrazide
9723
N,N'-bis(cyclohexylideneamino)oxamide
C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1
370-81-0
C14H22N4O2
278.356
MFCD00001659
DSRJIHMZAQEUJV-UHFFFAOYSA-N
cuprizone,cuprizane,biscyclohexanone oxaldihydrazone,ethanedioic acid, bis cyclohexylidenehydrazide,oxalic acid bis cyclohexylidenehydrazide,biscyclohexanone oxalyldihydrazone,bis cyclohexanone oxaldihydrazone,cuprizon,unii-5n16u7e0ao,oxalic acid, bis cyclohexylidenehydrazide
9723
N,N'-bis(cyclohexylideneamino)oxamide
C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1
Color White
Infrared Spectrum Authentic
Quantity 5g
Suitability suitable for determination of copper
Sulfated Ash 0.1% max.
Loss on Drying 0.5% max.
Melting Point 208°C to 214°C
Assay Percent Range 98%
Beilstein 07, IV, 25
Formula Weight 278.34
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Oxalic acid bis(cyclohexylidenehydrazide), 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-729-2

RTECSNumber : RO2520000

TSCA : TSCA