Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic acid dihydrate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
5kg
Packaging:
Plastic Bottle
Plastic drum
6153-56-6
C2H6O6
126.064
MFCD00149102
GEVPUGOOGXGPIO-UHFFFAOYSA-N
oxalic acid dihydrate,ethanedioic acid, dihydrate,unii-0k2l2ij59o,ethanedioic acid dihydrate,oxalic acid, dihydrate,oxalicacid dihydrate,dihydrate oxalic acid,oxalic acid di-hydrate,pubchem17416,acmc-209mtm
61373
oxalic acid;dihydrate
C(=O)(C(=O)O)O.O.O
6153-56-6
C2H6O6
126.064
MFCD00149102
GEVPUGOOGXGPIO-UHFFFAOYSA-N
oxalic acid dihydrate,ethanedioic acid, dihydrate,unii-0k2l2ij59o,ethanedioic acid dihydrate,oxalic acid, dihydrate,oxalicacid dihydrate,dihydrate oxalic acid,oxalic acid di-hydrate,pubchem17416,acmc-209mtm
61373
oxalic acid;dihydrate
C(=O)(C(=O)O)O.O.O
Assay Percent Range 99+%
Linear Formula HO2CCO2H·2H2O
Beilstein 02, 502
Fieser 01,764; 02,301; 05,481
Merck Index 15, 7010
Formula Weight 126.04
Physical Form Low Melting Crystalline Mass, Powder, Crystals and/or Chunks
Percent Purity ≥99%
Grade Extra Pure
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Packaging Plastic Bottle
Solubility Solubility in water: 138g/L (20°C). Other solubilities: 1200g/L in water (90°C), 400g/L in ethanol, 555g/L in boiling alcohol, 181g/L in glycerol, 10g/L in ether
Sulfate 1000ppm max.
Sulfated Ash 0.1% max.
Sublimation Point 157 °C
Melting Point 98.0°C to 102.0°C
Color Undesignated
Quantity 100g
Chemical Name or Material Oxalic acid dihydrate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger