Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

6153-56-6
C2H6O6
126.064
GEVPUGOOGXGPIO-UHFFFAOYSA-N
61373

High quality chemicals for laboratory and specialized industrial use

SMILES C(=O)(C(=O)O)O.O.O
IUPAC Name oxalic acid;dihydrate
Formula Weight 126.07
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.65kg
Quantity 2.5kg
Chemical Name or Material Oxalic Acid, Dihydrate
Grade Baker Analyzed™ ACS Reagent