Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic acid, potassium salt dihydrate, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Glass bottle
6100-20-5
C4H3KO8·2H2O
254.2
MFCD00150443
KPFHSTZUXXJAES-UHFFFAOYSA-M
potassium trihydrogen dioxalate dihydrate,potassium ion oxalic acid dihydrate hydrogen oxalate,potassium oxalic acid dihydrate hydrogen oxalate
131698588
potassium;hydron;oxalate;dihydrate
[H+].[H+].[H+].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.[K+]
6100-20-5
C4H3KO8·2H2O
254.2
MFCD00150443
KPFHSTZUXXJAES-UHFFFAOYSA-M
potassium trihydrogen dioxalate dihydrate,potassium ion oxalic acid dihydrate hydrogen oxalate,potassium oxalic acid dihydrate hydrogen oxalate
131698588
potassium;hydron;oxalate;dihydrate
[H+].[H+].[H+].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.O.[K+]
Packaging Glass bottle
Water 16% max. (K.F.)
Infrared Spectrum Authentic
Quantity 100g
Loss on Drying 12 % to 16 % (1 g, 105°C)
Color White to Beige
Assay Percent Range 99%
Merck Index 15, 7806
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in benzene, ethanol
Formula Weight 254.2
Physical Form Crystalline Powder or Solid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Oxalic acid, potassium salt dihydrate, 99%
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning