Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalyl bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
15219-34-8
C2Br2O2
215.83
MFCD00000113
JAZLVNXWYDFQFE-UHFFFAOYSA-N
oxalyl bromide,ethanedioyl dibromide,ethanedioyl bromide,oxalic dibromide,brcocobr,oxalyl bromide solution,ethanedioyl dibromide solution,oxalyl bromide solution, 2.0 m in methylene chloride
84841
oxalyl dibromide
C(=O)(C(=O)Br)Br
15219-34-8
C2Br2O2
215.83
MFCD00000113
JAZLVNXWYDFQFE-UHFFFAOYSA-N
oxalyl bromide,ethanedioyl dibromide,ethanedioyl bromide,oxalic dibromide,brcocobr,oxalyl bromide solution,ethanedioyl dibromide solution,oxalyl bromide solution, 2.0 m in methylene chloride
84841
oxalyl dibromide
C(=O)(C(=O)Br)Br
C-X bond formation reagent, Oxidation reagent
Specific Gravity 2.3
Melting Point -19.0°C
Refractive Index 1.5205 to 1.5225
Color Undesignated
Density 2.3000g/mL
Infrared Spectrum Authentic
Boiling Point 102.0°C to 103.0°C (720.0 mmHg)
Packaging Glass bottle
Quantity 25g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula BrCOCOBr
Fieser 01,767; 01,1137
Solubility Information Solubility in water: reacts with water.
Formula Weight 215.83
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Oxalyl bromide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 239-271-7