Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Glass bottle
79-37-8,75-44-5,76-02-8
C2Cl2O2
126.93
MFCD00000704
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
65578
oxalyl dichloride
C(=O)(C(=O)Cl)Cl
79-37-8,75-44-5,76-02-8
C2Cl2O2
126.93
MFCD00000704
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl chloride,ethanedioyl dichloride,oxalic dichloride,oxaloyl chloride,oxalic acid dichloride,oxalic acid chloride,ethanedioyl chloride,oxalylchloride,unii-r4y96317dw,cocl 2
65578
oxalyl dichloride
C(=O)(C(=O)Cl)Cl
C-X bond formation reagent, Oxidation reagent
Refractive Index 1.4285 to 1.4305
Specific Gravity 1.478
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Flash Point >100°C
Density 1.4780g/mL
Boiling Point 63.0°C to 64.0°C (763.0mmHg)
Color Colorless to Yellow
Melting Point -12.0°C
Assay Percent Range 97.5% min. (GC)
Linear Formula ClCOCOCl
Beilstein 02, 542
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Merck Index 15, 7013
Solubility Information Solubility in water: reacts. Other solubilities: soluble in the main common organic solvents :, methylene chloride; dichloroethane; chlorobenzene;, ethylacetate; hexane; acetone; toluene; ether;, benzene; chloroform and ether
Formula Weight 126.93
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Oxalyl chloride
Show More Show Less

GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Contact with w

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF INHA

GHS Signal Word: Danger

EINECSNumber : 201-200-2

RTECSNumber : KI2950000

TSCA : TSCA