Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxamic Hydrazide 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
515-96-8
C2H5N3O2
103.08
MFCD00008004
MOKRDWKSHLLYKM-UHFFFAOYSA-N
oxamic hydrazide,semioxamazide,semioxamazid,aminooxamide,oxamic acid hydrazide,oxamichydrazide,n-aminooxamide,1-hydrazinecarbonyl formamide,oxamic acid, hydrazide,unii-bet2ubd6nv
68200
2-hydrazinyl-2-oxoacetamide
C(=O)(C(=O)NN)N
515-96-8
C2H5N3O2
103.08
MFCD00008004
MOKRDWKSHLLYKM-UHFFFAOYSA-N
oxamic hydrazide,semioxamazide,semioxamazid,aminooxamide,oxamic acid hydrazide,oxamichydrazide,n-aminooxamide,1-hydrazinecarbonyl formamide,oxamic acid, hydrazide,unii-bet2ubd6nv
68200
2-hydrazinyl-2-oxoacetamide
C(=O)(C(=O)NN)N
Quantity 100g
Packaging Plastic bottle
Infrared Spectrum Authentic
Color White
Melting Point 216.0°C to 218.0°C
Assay Percent Range 99%
Linear Formula H2NCOCONHNH2
Beilstein 02, 559
Merck Index 15, 8584
Formula Weight 103.08
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Oxamic hydrazide, 99%
Show More Show Less

EINECSNumber : 208-213-2

RTECSNumber : AF2620000