Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxamide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
Plastic bottle
471-46-5
C2H4N2O2
88.066
MFCD00008007
YIKSCQDJHCMVMK-UHFFFAOYSA-N
ethanediamide,oxalamide,oxamimidic acid,oxalic acid diamide,diaminoglyoxal,oxamid,amid kyseliny stavelove,formimidic acid, 1-carbamoyl,oxamid czech,unii-sbe4m0223e
10113
CHEBI:48248
oxamide
C(=O)(C(=O)N)N
471-46-5
C2H4N2O2
88.066
MFCD00008007
YIKSCQDJHCMVMK-UHFFFAOYSA-N
ethanediamide,oxalamide,oxamimidic acid,oxalic acid diamide,diaminoglyoxal,oxamid,amid kyseliny stavelove,formimidic acid, 1-carbamoyl,oxamid czech,unii-sbe4m0223e
10113
CHEBI:48248
oxamide
C(=O)(C(=O)N)N
Color White
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NCOCONH2
Beilstein 02, 545
Merck Index 15, 7017
Formula Weight 88.06
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Oxamide
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.