Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxazepam Polyclonal Antibody, Invitrogen™

Catalog No. PIPA175328
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA175328 100 μL
1 options

Catalog No. PIPA175328

Supplier: Thermo Scientific PA175328

Sheep Polyclonal Antibody

PA1-75328 specifically detects Oxazepam. PA1-75328 has been successfully used in ELISA applications. The PA1-75328 immunogen is Oxazepam(N)-BSA.

Oxazepam is only found in individuals that have used or taken this drug. It is an intermediate-acting benzodiazepine used to treat alcohol withdrawal and anxiety disorders. Similar to other benzodiazepines, oxazepam exerts its anxiolytic effects by potentiating the effect of gamma-aminobutyric acid (GABA) on GABA-A receptors through a cooperative mechanism of action. GABA receptors are ionotropic chloride-linked channel receptors that produce inhibitory postsynaptic potentials. When activated by GABA, the GABA receptor/chloride ionophore complex undergoes a conformational change that allows the passage of chloride ions through the channel. Benzodiazepines are believed to exert their effect by increasing the effect of GABA at its receptor. Benzodiazepine binding increases chloride conductance in the presence of GABA by increasing the frequency at which the channel opens. In contrast, barbiturates increase chloride conductance in the presence of GABA by prolonging the time in which the channel remains open. There are 18 subtypes of the GABA receptor subunits. The a2 subunit of the alpha-2, beta-3, gamma-2 receptor complex is thought to mediate anxiolytic effects while the a1 subunit of the alpha-1, beta-2, gamma-2 receptor complex is thought to mediate sedative, anticonvulsant and anterograde amnesia effects. [National Center for Biotechnology Information. PubChem Compound Database; CID=4616]
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oxazepam
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Gene Alias Tazepam; Serax; Adumbran; Droxacepam; Durazepam; C15H11ClN2O2
Host Species Sheep
Immunogen Oxazepam(N)-BSA
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.