Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxindole 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
59-48-3
C8H7NO
133.15
MFCD00005711
JYGFTBXVXVMTGB-UHFFFAOYSA-N
oxindole,indolin-2-one,2-oxindole,2-indolinone,1,3-dihydro-2h-indol-2-one,oxindol,2-oxoindoline,2-oxindoline,indol-2 3h-one,2h-indol-2-one, 1,3-dihydro
321710
CHEBI:31697
1,3-dihydroindol-2-one
C1C2=CC=CC=C2NC1=O
59-48-3
C8H7NO
133.15
MFCD00005711
JYGFTBXVXVMTGB-UHFFFAOYSA-N
oxindole,indolin-2-one,2-oxindole,2-indolinone,1,3-dihydro-2h-indol-2-one,oxindol,2-oxoindoline,2-oxindoline,indol-2 3h-one,2h-indol-2-one, 1,3-dihydro
321710
CHEBI:31697
1,3-dihydroindol-2-one
C1C2=CC=CC=C2NC1=O
Melting Point 123°C to 128°C
Boiling Point 227°C (73.0mmHg)
Quantity 25g
Packaging Glass bottle
Assay Percent Range 97%
Beilstein 21, 282
Solubility Information Solubility in water: insoluble. Other solubilities: soluble inn most organic solvents
Formula Weight 133.15
Grade Pure
Chemical Name or Material Oxindole, 97%

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 200-429-5

RTECSNumber : NM2080500