Learn More
Thermo Scientific™ Oxoid™ C-T Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.
$368.55 - $796.43
Specifications
Description C-T Supplement
Format Freeze Dried
Packaging 10/Pk.
Quantity 10 x Freeze Dried Vials
View More Specs
Products 2
Catalog Number Mfr. No. Yield Price Quantity & Availability  
Catalog Number Mfr. No. Yield Price Quantity & Availability  
OXSR0172E Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
SR0172E
Each vial for 500mL medium Pack of 10 for $368.55
Add to cart
OXSR0172H Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
SR0172H
Each vial for 2L medium Pack of 10 for $796.43
Add to cart
Description
Description
Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.
Add to Sorbitol MacConkey Agar, Part No. CM0813B or to Sorbitol MacConkey Agar with BCIG, Part No. CM0981B.
Ordering Information:
Each vial supplements 2L of medium.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
C-T Supplement
Freeze Dried
10/Pk.
10 x Freeze Dried Vials