Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ CAT Supplement

The Oxoid CAT Supplement isolates Campylobacter spp.

Manufacturer:  Thermo Scientific™ SR0174H

 View more versions of this product

Catalog No. SR0174H


Add to cart
This item is not returnable. View return policy

Description

Description

Oxoid CAT Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Acrobacter.

  • Add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B for the isolation of Campylobacter spp.
  • Add to Arcobacter Broth Base, Part No. CM0965B for the isolation of Arcobacter spp.
Specifications

Specifications

Cat Sel Suppl For 2l 10/Pk.
Freeze Dried
For 2L medium
Supplement
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit