Learn More
Thermo Scientific™ Oxoid™ CAT Supplement Available on GSA/VA Contract for Federal Government customers only.
The Oxoid CAT Supplement isolates Campylobacter spp.
$286.13 - $758.23
Specifications
Type Supplement
Format Freeze Dried
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SR0174H
View Documents
Thermo Scientific™
SR0174H
For 2L medium Pack of 10 for $758.23
Add to cart
OXSR0174E Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
SR0174E
10 x Freeze Dried Vials Pack of 10 for $286.13
Add to cart
Description
Description
Oxoid CAT Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Acrobacter.
  • Add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B for the isolation of Campylobacter spp.
  • Add to Arcobacter Broth Base, Part No. CM0965B for the isolation of Arcobacter spp.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Supplement
Freeze Dried