Learn More
Thermo Scientific™ Oxoid™ Gardnerella Vaginalis Selective Supplement
The Oxoid Gardnerella Vaginalis Selective Supplement is used for the isolation of Gardnerella vaginalis.
Manufacturer:  Thermo Scientific™ SR0119E
Catalog No. SR0119E
Add to cart
This item is not returnable. View return policy
Description
Description
Oxoid Gardnerella Vaginalis Selective Supplement is used for the isolation of Gardnerella vaginalis.
  • Add to to Columbia Blood Agar Base, Part No. CM0331B
  • Each vial supplements 500mL of medium
Specifications
Specifications
Gardnerella Vaginalis Selective Supplement
10/Pk.
Supplement
Freeze Dried
10 x Freeze Dried Vials
Each vial for 500mL medium