Learn More
Thermo Scientific™ Oxoid™ Legionella BCYE Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.
$253.05 - $894.60
Specifications
Description Legionella BCYE Supplement
Format Freeze Dried
Packaging 10/Pk.
Quantity 10 x Freeze Dried Vials
View More Specs
Products 2
Catalog Number Mfr. No. Yield Price Quantity & Availability  
Catalog Number Mfr. No. Yield Price Quantity & Availability  
OXSR0110A Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
SR0110A
Each vial for 100mL medium
Pack of 10 for $253.05
Add to cart
OXSR0110C Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
SR0110C
Each vial for 500mL medium
Pack of 10 for $894.60
Add to cart
Description
Description
Oxoid Legionella BCYE Supplement is used for the isolation of Legionella spp.
Add to Legionella CYE Agar Base, Part No. CM0655B
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Legionella BCYE Supplement
Freeze Dried
10/Pk.
10 x Freeze Dried Vials