Learn More
Thermo Scientific™ Oxoid™ Listeria Selective Enrichment Supplement Available on GSA/VA Contract for Federal Government customers only.
Oxoid Listeria Selective Enrichment Supplement is used for the enrichment of Listeria spp.
Manufacturer:  Thermo Scientific™ SR0141E
Catalog No. OXSR0141E
Add to cart
Description
Description
Oxoid Listeria Selective Enrichment Supplement is used for the enrichment of Listeria spp.
  • Add to Listeria Enrichment Broth, Part No. CM0862B
  • Each vial supplements 500mL of medium
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
10 x Freeze Dried Vials
Freeze Dried
Supplement
10/Pk.
Listeria Selective Enrichment Supplement
Each vial for 500mL medium