Learn More
Thermo Scientific™ Oxoid™ Modified Listeria Selective Supplement
The Oxoid Listeria Selective Supplement, Modified (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Manufacturer:  Thermo Scientific™ SR0213E
Catalog No. SR0213E
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Oxoid Listeria Selective Supplement, (Modified Listeria Selective Supplement) is an alternative supplement containing amphotericin B as a replacement for cycloheximide.
Add to to Buffered Listeria Enrichment Broth, Part No. CM0897R or Listeria Enrichment Broth, Part No. CM0862B. For the enrichment of Listeria monocytogenes. Each vial supplements 500mL of medium.
Specifications
Specifications
Listeria Selective Supplement, Modified
10/Pk.
Supplement
Freeze Dried
10 x Freeze Dried Vials
Each vial for 500mL medium