Learn More
Thermo Scientific™ Oxoid™ Oxford Listeria Selective Supplement
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Manufacturer:  Thermo Scientific™ SR0206B
 View more versions of this product
Catalog No. SR0206B
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
  • Add to Listeria Selective Agar Base, Part No. CM0856B
Specifications
Specifications
Mod Lister(oxf)sup For 2.5L 10Pk.
For 2.5L medium