Learn More
Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using this test kit.
Supplier:  Thermo Scientific™ DR0591G
 View more versions of this product
Catalog No. OXDR0591G
Add to Cart
This item is not returnable. View return policy
Description
Description
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using Thermo Scientific™ Oxoid™ Streptococcal Grouping Kit. This rapid latex agglutination test is designed to deliver simple, rapid and sensitive results for the identification of streptococcal groups A, B, C, D, F, and G . The test utilizes latex particles coated with group-specific antibodies which will agglutinate in the presence of homologous antigens.
  • Supplied with rapid latex agglutination test
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Streptococcal Grouping Reagent G
Reagents for Streptococcus Testing
Group A, B, C, D, F and G
50 Tests/Vial