Learn More
Thermo Scientific™ Oxoid™ UVMII Listeria Secondary Selective Enrichment Supplement
The Oxoid Listeria Secondary Selective Enrichment Supplement, UVMII is used in a two-step enrichment method for enrichment of Listeria spp.
Manufacturer:  Thermo Scientific™ SR0143E
Catalog No. SR0143E
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Oxoid Listeria Secondary Selective Enrichment Supplement, UVMII is used in a two-step selective enrichment (USDS-FSIS) method for the enrichment of Listeria spp.
  • Add to Listeria Enrichment Broth Base (UVM formulation), Part No. CM0863B
  • Each vial supplements 500mL of medium
Specifications
Specifications
Listeria Secondary Selective Enrichment Supplement, UVMII
10/Pk.
Supplement
Freeze Dried
10 x Freeze Dried Vials
Each vial for 500mL medium