Learn More
Thermo Scientific™ Oxoid™ Vancomycin Supplement
Used for addition to selective media for isolation of Vancomycin Resistant Enterococci and High Level Aminoglycoside Resistant Enterococci from clinical specimens.
$116.87 - $299.25
Specifications
Description Oxoid™ Vancomycin Supplement
Quantity 10/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
SR0247E
View Documents
Thermo Scientific™
SR0247E
Powder 10/Pk.
Pack of 10 for $299.25
Add to cart
OXSR0186E
View Documents
Thermo Scientific™
SR0186E
Freeze-dried 10/Pk.
Pack of 10 for $116.87
Add to cart
Description
Description
Thermo Scientific™ Oxoid Vancomycin Supplement is for addition to selective media for the isolation of Vancomycin Resistant Enterococci (VRE) and High Level Aminoglycoside Resistant Enterococci (HLARE) from clinical specimens.
  • Each vial supplements 500mL of medium
Specifications
Specifications
Oxoid™ Vancomycin Supplement
10/Pk.