Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Oxsorbent, Gas Analysis Chemcials, Burrell Scientific

Determining oxygen levels in gas analysis

Manufacturer:  Burrell Scientific Inc 0397100008

Catalog No. 14-400-151



Description

Description

  • For Use with Gas Analyzers
  • Product for determining oxygen levels in gas analysis
  • Fast, clean, and accurate solution for testing
  • Can absorb up to four times its volume in oxygen

Recommended Storage

Store at room temperature

Warranty and Services

6 Month Limited Warranty

Specifications

Specifications

Oxsorbent, Gas Analysis Chemcials
10049-05-5, 7647-01-0
Liquid
8 oz.
73.50%, 25%, 1.50%
>100°C (>212°F)
Blue
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit