Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXSR1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548667

Catalog No. PA548667

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.
Specifications

Specifications

Oxsr1
Polyclonal
PBS with 0.09% sodium azide
O95747, Q6P9R2
Serine/threonine-protein kinase OSR1, Oxidative stress-responsive 1 protein, Oxsr1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
108737, 9943
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
OXSR1
Liquid
OXSR1
KLH conjugated synthetic peptide between 94-121 amino acids from the N-terminal region of mouse Oxsr1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit