Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxychlordane, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
27304-13-8,67-56-1
C10H4Cl8O
423.741
VWGNQYSIWFHEQU-IBAUKUMTSA-N
46174053
CHEBI:81541
C12C(C(C3(C1O3)Cl)Cl)C4(C(=C(C2(C4(Cl)Cl)Cl)Cl)Cl)Cl
27304-13-8,67-56-1
C10H4Cl8O
423.741
VWGNQYSIWFHEQU-IBAUKUMTSA-N
46174053
CHEBI:81541
C12C(C(C3(C1O3)Cl)Cl)C4(C(=C(C2(C4(Cl)Cl)Cl)Cl)Cl)Cl

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Oxychlordane27304-13-81000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
CAS Min % 0.1,99.9
CAS Max % 0.1,99.9
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Solvent or Matrix Methanol
Quantity 1mL
Chemical Name or Material Oxychlordane

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only