Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxymetazoline hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
2321-07-5
C20H12O5
332.31
GNBHRKFJIUUOQI-UHFFFAOYSA-N
fluorescein,solvent yellow 94,resorcinolphthalein,yellow fluorescein,3,6-fluorandiol,d and c yellow no. 7,fluoresceine,japan yellow 201,c.i. solvent yellow 94,FD&C yellow no. 7
16850
CHEBI:31624
3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O
2321-07-5
C20H12O5
332.31
GNBHRKFJIUUOQI-UHFFFAOYSA-N
fluorescein,solvent yellow 94,resorcinolphthalein,yellow fluorescein,3,6-fluorandiol,d and c yellow no. 7,fluoresceine,japan yellow 201,c.i. solvent yellow 94,FD&C yellow no. 7
16850
CHEBI:31624
3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

α1A agonist

Partial α 2A adrenoceptor agonist and α 1A agonist. Agonist/antagonist activity at 5-HT receptors.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 296.84
Percent Purity >98%
Chemical Name or Material Oxymetazoline hydrochloride

Recommended Storage : Store at Room Temperature