Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oxyntomodulin Mouse anti-Human, Mouse, Rat, Clone: 20, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Oxyntomodulin |
---|---|
Clone | 20 |
Concentration | 1 mg/mL |
Dilution | ELISA |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP26194501
|
Novus Biologicals
NBP2619450.1ML |
0.1 mL |
Each for $436.00
|
|
NB019771
|
Novus Biologicals
NBP2619450.025ML |
0.025 mL |
Each for $176.85
|
|
Description
Oxyntomodulin Monoclonal specifically detects Oxyntomodulin in Human samples. It is validated for ELISA.Specifications
Oxyntomodulin | |
1 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
0.01 M phosphate buffer, pH 7.4, containing 0.5 M NaCl with 15mM Sodium Azide | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
20 | |
ELISA | |
Monoclonal | |
Purified | |
Human, Mouse, Rat | |
Synthetic full length oxyntomodulin coupled to carrier | |
Protein A purified | |
Oxyntomodulin Antibody is specific for human oxyntomodulin. Cross-reacts fully with glucagon. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title