Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Oxytetracycline hydrochloride, 50mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
25mL
50mL
2058-46-0
C22H25ClN2O9
496.897
MFCD00135815
UTAHTGPLLFLKGW-IFLJXUKPSA-N
121235537
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl
2058-46-0
C22H25ClN2O9
496.897
MFCD00135815
UTAHTGPLLFLKGW-IFLJXUKPSA-N
121235537
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl
Physical Form Liquid
Quantity 25mL
Sensitivity Light Sensitive
Chemical Name or Material Oxytetracycline hydrochloride, 50mg/mL in distilled water, sterile-filtered

RTECSNumber : QI8225000

TSCA : TSCA

storageNote1 : -30° to -10°C