Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ozagrel hydrochloride, Tocris Bioscience™

Catalog Number 151050
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
74003-18-2
C13H13ClN2O2
264.709
CWKFWBJJNNPGAM-IPZCTEOASA-N
e-3-4-1h-imidazol-1-yl methyl phenyl acrylic acid hydrochloride, 3-4-imidazol-1-ylmethyl phenyl prop-2-enoic acid hydrochloride
6438130
CHEBI:31954
(E)-3-[4-(imidazol-1-ylmethyl)phenyl]prop-2-enoic acid;hydrochloride
C1=CC(=CC=C1CN2C=CN=C2)C=CC(=O)O.Cl
This item is not returnable. View return policy
74003-18-2
C13H13ClN2O2
264.709
CWKFWBJJNNPGAM-IPZCTEOASA-N
e-3-4-1h-imidazol-1-yl methyl phenyl acrylic acid hydrochloride, 3-4-imidazol-1-ylmethyl phenyl prop-2-enoic acid hydrochloride
6438130
CHEBI:31954
(E)-3-[4-(imidazol-1-ylmethyl)phenyl]prop-2-enoic acid;hydrochloride
C1=CC(=CC=C1CN2C=CN=C2)C=CC(=O)O.Cl

Selective thromboxane A2 synthetase inhibitor

Potent and selective inhibitor of thromboxane (TXA2) synthetase (IC50 = 4 nM). Does not inhibit prostacyclin (PGI2) synthetase, cyclooxygenase or PGE2 isomerase (IC50 > 1 mM). Inhibits platelet aggregation in vitro and arachidonate-induced arterial contraction in vivo.
Quantity 50 mg
Formula Weight 264.71
Percent Purity >98%
Chemical Name or Material Ozagrel hydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit