Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ozark River Coated Wire Mounting Glove Dispenser

For use with Ozark River Portable Hygienic Hand-Washing Stations

Manufacturer:  Ozark River AC07GLV

Catalog No. 22-535-019

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Durable coated wire glove box holder can be installed onto a wall or on Ozark River Portable Hygienic Hand-Washing Stations
  • Holds one box horizontally or two boxes vertically

Order Info

Fits glove boxes 9.75W x 5.25D x 3.25 in. H (24.8 x 13.3 x 8.25cm)

Specifications

Specifications

5.25 in.
Ozark river portable hygienic hand-washing stations
Glove Dispenser
8.25cm
539.7mm
13.3cm
1 horizontal Box and 2 vertical boxes
3.25 in.
12.5 in.
Coated Wire
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit