Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisaldehyde dimethyl acetal, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
2186-92-7
C10H14O3
182.22
MFCD00036507
NNHYAHOTXLASEA-UHFFFAOYSA-N
1-dimethoxymethyl-4-methoxybenzene,p-anisaldehyde dimethyl acetal,anisaldehyde dimethyl acetal,4-methoxybenzaldehyde dimethyl acetal,p-dimethoxymethyl anisole,benzene, 1-dimethoxymethyl-4-methoxy,p-anisaldehydedimethylacetal,unii-xte77a5et4,anisicaldehyde dimethylacetal,p-anisaldehyde dimethylacetal
75140
1-(dimethoxymethyl)-4-methoxybenzene
COC1=CC=C(C=C1)C(OC)OC
2186-92-7
C10H14O3
182.22
MFCD00036507
NNHYAHOTXLASEA-UHFFFAOYSA-N
1-dimethoxymethyl-4-methoxybenzene,p-anisaldehyde dimethyl acetal,anisaldehyde dimethyl acetal,4-methoxybenzaldehyde dimethyl acetal,p-dimethoxymethyl anisole,benzene, 1-dimethoxymethyl-4-methoxy,p-anisaldehydedimethylacetal,unii-xte77a5et4,anisicaldehyde dimethylacetal,p-anisaldehyde dimethylacetal
75140
1-(dimethoxymethyl)-4-methoxybenzene
COC1=CC=C(C=C1)C(OC)OC
Color Colorless to Yellow
Boiling Point 235.0°C
Packaging Glass bottle
Flash Point >110°C
Quantity 500g
Density 1.0680g/mL
Refractive Index 1.5045 to 1.5065
Infrared Spectrum Authentic
Specific Gravity 1.068
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,soluble in common organic solvents
Formula Weight 182.22
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material p-Anisaldehyde dimethyl acetal
Show More Show Less