Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
100-09-4
C8H8O3
152.15
MFCD00002542
ZEYHEAKUIGZSGI-UHFFFAOYSA-N
p-anisic acid,anisic acid,p-methoxybenzoic acid,draconic acid,4-anisic acid,benzoic acid, 4-methoxy,anisic acid, para,4-methoxybenzoate,para-anisic acid,anisic acid, p-isomer
7478
CHEBI:40813
4-methoxybenzoic acid
COC1=CC=C(C=C1)C(=O)O
100-09-4
C8H8O3
152.15
MFCD00002542
ZEYHEAKUIGZSGI-UHFFFAOYSA-N
p-anisic acid,anisic acid,p-methoxybenzoic acid,draconic acid,4-anisic acid,benzoic acid, 4-methoxy,anisic acid, para,4-methoxybenzoate,para-anisic acid,anisic acid, p-isomer
7478
CHEBI:40813
4-methoxybenzoic acid
COC1=CC=C(C=C1)C(=O)O
Quantity 1kg
Boiling Point 275.0°C
Infrared Spectrum Authentic
Melting Point 181.0°C to 186.0°C
Refractive Index 1.5710 to 1.5760
Flash Point 185°C
Color Beige-Gray to White
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula CH3OC6H4CO2H
Beilstein 10,IV,346
Merck Index 15,659
Solubility Information Solubility in water: 0.3g/L (20 c). Other solubilities: freely soluble in alcohol,ether,chloroform and,ethyl acetate
Formula Weight 152.15
Physical Form Powder
Percent Purity 98%
Chemical Name or Material p-Anisic acid, 98%
Show More Show Less

EINECSNumber : 202-818-5

RTECSNumber : BZ4395000

TSCA : TSCA