Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisidine-3-sulfonic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
6470-17-3
C7H9NO4S
203.212
MFCD00035760
JXZGTFLJFKLVAX-UHFFFAOYSA-N
4-Aminoanisole-2-sulfonic Acid, 3-Amino-6-methoxybenzenesulfonic Acid, 4-Methoxyaniline-3-sulfonic Acid
80945
5-amino-2-methoxybenzenesulfonic acid
COC1=C(C=C(C=C1)N)S(=O)(=O)O
6470-17-3
C7H9NO4S
203.212
MFCD00035760
JXZGTFLJFKLVAX-UHFFFAOYSA-N
4-Aminoanisole-2-sulfonic Acid, 3-Amino-6-methoxybenzenesulfonic Acid, 4-Methoxyaniline-3-sulfonic Acid
80945
5-amino-2-methoxybenzenesulfonic acid
COC1=C(C=C(C=C1)N)S(=O)(=O)O
Quantity 25g
Color Gray-White
Formula Weight 203.21
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material p-Anisidine-3-sulfonic Acid

EINECSNumber : (3)-2039

TSCA : Yes