Alfa Aesar™ p-Anisidine, 99%

Manufacturer: Alfa Aesar™ A1094636

Quantity 500g

 View more versions of this product

Catalog No. AAA1094636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104-94-9
    Molecular Formula C7H9NO
    Formula Weight 123.16
    MDL Number MFCD00007864
    UN Number UN2811
    Beilstein 471556
    Merck Index 14,667
    Synonym 4-Aminoanisole; 4-Methoxyaniline
    Chemical Name or Material p-Anisidine

    Specifications

    Formula Weight 123.16
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    TSCA TSCA
    UN Number UN2811
    Boiling Point 240° to 243°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    RTECS Number BZ5450000
    Molecular Formula C7H9NO
    MDL Number MFCD00007864
    CAS 104-94-9
    Beilstein 471556
    Merck Index 14,667
    Synonym 4-Aminoanisole; 4-Methoxyaniline
    Quantity 500g
    Flash Point 122°C (251°F)
    Melting Point 56° to 60°C
    Chemical Name or Material p-Anisidine
    Assay 99%

    Description

    Applications
    p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

    Solubility
    Soluble in acetone, benzene, ether and ethanol. Insoluble in water.

    Notes
    Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.

    GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit