Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisidine 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
104-94-9
C7H9NO
123.155
MFCD00007864
BHAAPTBBJKJZER-UHFFFAOYSA-N
p-anisidine,4-anisidine,4-methoxybenzenamine,4-aminoanisole,p-aminoanisole,para-anisidine,p-methoxyaniline,benzenamine, 4-methoxy,4-methoxybenzeneamine,p-anisylamine
7732
CHEBI:82388
4-methoxyaniline
COC1=CC=C(C=C1)N
104-94-9
C7H9NO
123.155
MFCD00007864
BHAAPTBBJKJZER-UHFFFAOYSA-N
p-anisidine,4-anisidine,4-methoxybenzenamine,4-aminoanisole,p-aminoanisole,para-anisidine,p-methoxyaniline,benzenamine, 4-methoxy,4-methoxybenzeneamine,p-anisylamine
7732
CHEBI:82388
4-methoxyaniline
COC1=CC=C(C=C1)N
Quantity 500g
Melting Point 59°C
Boiling Point 246°C
Color Undesignated
UN Number 2811
Formula Weight 123.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material p-Anisidine

EINECSNumber : (3)-0682

RTECSNumber : BZ5450000

TSCA : Yes