Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Anisylchlorodiphenylmethane, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
14470-28-1
C20H17ClO
308.8
MFCD00000814
OBOHMJWDFPBPKD-UHFFFAOYSA-N
4-methoxytriphenylchloromethane,4-methoxytrityl chloride,4-methoxytriphenylmethyl chloride,4-monomethoxytrityl chloride,mmtrcl,p-chlorodiphenylmethyl anisole,p-anisylchlorodiphenylmethane,1-chlorodiphenylmethyl-4-methoxybenzene,chloro 4-methoxyphenyl methylene dibenzene,4-methoxytritylchloride
84462
1-[chloro(diphenyl)methyl]-4-methoxybenzene
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
14470-28-1
C20H17ClO
308.8
MFCD00000814
OBOHMJWDFPBPKD-UHFFFAOYSA-N
4-methoxytriphenylchloromethane,4-methoxytrityl chloride,4-methoxytriphenylmethyl chloride,4-monomethoxytrityl chloride,mmtrcl,p-chlorodiphenylmethyl anisole,p-anisylchlorodiphenylmethane,1-chlorodiphenylmethyl-4-methoxybenzene,chloro 4-methoxyphenyl methylene dibenzene,4-methoxytritylchloride
84462
1-[chloro(diphenyl)methyl]-4-methoxybenzene
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
Quantity 100g
Packaging Glass bottle
Melting Point 121.0°C to 125.0°C
Assay Percent Range 96% min
Linear Formula CH3OC6H4C(C6H5)2Cl
Beilstein 06, 713
Formula Weight 308.8
Percent Purity 97%
Chemical Name or Material p-Anisylchlorodiphenylmethane, 97%

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Warning

EINECSNumber : 238-463-8