Alfa Aesar™ p-Arsanilic acid, 98%

Manufacturer: Alfa Aesar™ B2459536

Quantity 500g

 View more versions of this product

Catalog No. AAB2459536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98-50-0
    Molecular Formula C6H8AsNO3
    Formula Weight 217.06
    MDL Number MFCD00007819
    UN Number UN3465
    Beilstein 1102334
    Merck Index 14,792
    Synonym 4-Aminobenzenearsonic acid; 4-Aminophenylarsonic acid
    Chemical Name or Material p-Arsanilic acid

    Specifications

    Formula Weight 217.06
    CAS 98-50-0
    UN Number UN3465
    Beilstein 1102334
    Quantity 500g
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
    TSCA TSCA
    Molecular Formula C6H8AsNO3
    MDL Number MFCD00007819
    Merck Index 14,792
    Synonym 4-Aminobenzenearsonic acid; 4-Aminophenylarsonic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Melting Point >300°C
    RTECS Number CF7875000
    Chemical Name or Material p-Arsanilic acid
    Assay 98%

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit