Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ p-Arsanilic acid, 98%

Click to view available options
Quantity:
100g
500g
98-50-0
C6H8AsNO3
217.056
MFCD00007819
XKNKHVGWJDPIRJ-UHFFFAOYSA-N
p-arsanilic acid,arsanilic acid,4-arsanilic acid,4-aminophenyl arsonic acid,4-aminobenzenearsonic acid,atoxylic acid,p-aminobenzenearsonic acid,aminophenylarsine acid,p-anilinearsonic acid,pro-gen
7389
CHEBI:49477
(4-aminophenyl)arsonic acid
C1=CC(=CC=C1N)[As](=O)(O)O
98-50-0
C6H8AsNO3
217.056
MFCD00007819
XKNKHVGWJDPIRJ-UHFFFAOYSA-N
p-arsanilic acid,arsanilic acid,4-arsanilic acid,4-aminophenyl arsonic acid,4-aminobenzenearsonic acid,atoxylic acid,p-aminobenzenearsonic acid,aminophenylarsine acid,p-anilinearsonic acid,pro-gen
7389
CHEBI:49477
(4-aminophenyl)arsonic acid
C1=CC(=CC=C1N)[As](=O)(O)O
Melting Point >300°C
Quantity 100g
UN Number UN3465
Beilstein 1102334
Merck Index 14,792
Formula Weight 217.06
Percent Purity 98%
Chemical Name or Material p-Arsanilic acid

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.

RTECSNumber : CF7875000

TSCA : TSCA