Learn More
P-Cadherin Mouse anti-Human, Alexa Fluor 350, Clone: 104805R, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB861RU100UG
Catalog No. FAB861RU100
Add to cart
This item is not returnable. View return policy
Description
Description
P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
P-Cadherin
104805R
Flow Cytometry 0.25-1 μg/10^6 cells
CAA45177
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin
Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human P-Cadherin in direct ELISAs.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
1001
Human