Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human P-Cadherin Alexa Fluor 700-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB861RN100UG
This item is not returnable.
View return policy
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry.Specifications
P-Cadherin | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177 | |
100 μg | |
1001 | |
Human | |
Supernatant |
Flow Cytometry | |
104805R | |
Flow Cytometry 0.25-1 μg/10^6 cells | |
CAA45177 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human P-Cadherin in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction