Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Cadherin Mouse anti-Human, Clone: 12H6, Novus Biologicals™

Mouse Monoclonal Antibody

$152.22

Specifications

Antigen P-Cadherin
Clone 12H6
Host Species Mouse
Isotype IgG1 Kappa
Gene Alias cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25327920 View Documents Novus Biologicals
NBP25327920UG
20 ug Each for $152.22
Add to cart
 
Description

Description

P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

P-Cadherin
Mouse
cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
12H6
IgG1 Kappa
Human
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit