Learn More
P-Cadherin Rat anti-Mouse, Alexa Fluor 700, Clone: 106020, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB761N100UG
Catalog No. FAB761N100
Add to cart
This item is not returnable. View return policy
Description
Description
P-Cadherin Monoclonal antibody specifically detects P-Cadherin in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
P-Cadherin
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q8BSL6
CDH3
Mouse myeloma cell line NS0-derived recombinant mouse P-Cadherin Glu100-Gly647 Accession # Q8BSL6
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse P-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) N-Cadherin, recombinant mouse (rm) VE-Cadherin, rhCadherin-8, or rhCadherin-17 is observed.
Flow Cytometry
106020
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin
Rat
IgG2a
RUO
Primary
1001
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)