Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p-Dimethylamino-Benzaldehyde, ACS, MP Biomedicals™
Click to view available options
Quantity:
50 g
100 g
250 g
Specifications
Quantity | 50 g |
Formula Weight | 149.2 |
Grade | ACS Reagent |
Physical Form | Solid |
Chemical Name or Material | p-Dimethylamino-Benzaldehyde |
Safety and Handling
EINECSNumber | 202-819-0 |
storageNote1 | Store under nitrogen, protect from light. |
Recommended Storage | Store at Room Temperature (15deg to 30°C) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction