Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dimethylaminoazobenzene, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
60-11-7,67-56-1
C14H15N3
225.295
JCYPECIVGRXBMO-UHFFFAOYSA-N
6053
CHEBI:17903
N,N-dimethyl-4-phenyldiazenylaniline
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2
60-11-7,67-56-1
C14H15N3
225.295
JCYPECIVGRXBMO-UHFFFAOYSA-N
6053
CHEBI:17903
N,N-dimethyl-4-phenyldiazenylaniline
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
p-Dimethylaminoazobenzene60-11-71000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.1,99.9
CAS Max % 0.1,99.9
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Solvent or Matrix Methanol-P&T
Quantity 1mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Chemical Name or Material p-Dimethylaminoazobenzene

DOTInformation : Nonhazardous

Recommended Storage : Freezer

Product intended for laboratory use only