Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Dioxane, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
150mL
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

123-91-1
C4H8O2
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
CHEBI:47032
1,4-dioxane
C1COCCO1

High quality chemicals for laboratory and specialized industrial use

Flash Point 12.00°C
Quantity 1L
Packaging Aluminum Bottle
Specific Gravity 1L = 1.03kg
Formula Weight 88.11
Chemical Name or Material p-Dioxane
Grade Baker Analyzed™ ACS Reagent